missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%
Supplier: Honeywell-Fluka 332095L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Acetic acid | |
| C2H4O2 | |
| MFCD00036152 | |
| 506007 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05g/mol | |
| ACS/puriss. p.a. | |
| 5 L | |
| 40°C (104°F) | |
| Plastic Bottle |
| 64-19-7 | |
| CH3CO2H | |
| UN2789 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ≥99.8% | |
| 1.049g/mL at 25°C | |
| 16% at 92°F, 4% at 59°F | |
| 800°F | |
| 2.07 (vs air) |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
RTECSNumber : AF1225000