missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Solstice

Catalog No. 6000035
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 kg
30 kg
4 x 2.5 L
60 kg
6 x 1 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000035 60 kg
6000031 1 L
6000032 2.5 L
60045760 6 x 1 L
6000030 10 kg
60045759 4 x 2.5 L
6000033 30 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 6000035 Supplier Solstice Supplier No. 2722560KG
Only null left

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number 2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity 99.8 to 100.5%
Grade puriss.
Density 1.049 g/mL at 25°C
Quantity 60 kg
Explosion Limit 16% (92°F), 4% (59°F)
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Plastic Pail
Vapor Density 2.07 (vs. air)
Chemical Name or Material Acetic Acid, Meets analytical specification of Ph. Eur.BP, USP, FCC
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H226-H314
EINECSNumber 200-580-7
RTECSNumber AF1225000