missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss., 99-100%, Solstice

Catalog No. 60045757
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045757 6 x 1 L
6000027 1 L
6000028 2.5 L
6000029 5 L
60045755 4 x 2.5 L
60045756 4 x 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60045757 Supplier Solstice Supplier No. 272216X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss., 99-100%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.2°C (literature)
Boiling Point 118°C
CAS Max % 100.0000%
Density 1.049 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point 40°C
Packaging Poly Bottle
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Percent Purity >99%
Grade puriss.
Chemical Name or Material Acetic acid, Puriss., 99-100%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Acetic acid, glacial
EINECSNumber 200-580-7
Recommended Storage Room Temp