missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, pure, anhydrous
CAS: 127-08-2 | C2H3KO2 | 98.14 g/mol
Supplier: Thermo Scientific Chemicals 220150010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acetic acid, potassium salt | |
| 99+% | |
| White | |
| 100.0 | |
| 1 kg | |
| C2H3KO2 | |
| MFCD00012458 | |
| 01,906; 06,475 | |
| 15,7726 | |
| Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
| CC(=O)[O-].[K+] | |
| 98.14 | |
| CHEBI:32029 | |
| ≥99% | |
| Authentic |
| 292.0°C | |
| 127-08-2 | |
| 99.0 | |
| Granular Crystalline Powder | |
| 99+% | |
| CH3CO2K | |
| 02,96 | |
| >250°C | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.14 | |
| Pure | |
| Plastic bottle |
Chemical Identifiers
| 127-08-2 | |
| 98.14 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 204-822-2
RTECSNumber : AJ3325000
TSCA : TSCA
RUO – Research Use Only