missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, for molecular biology
CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
$55.47 - $55.47
Chemical Identifiers
| CAS | 127-08-2 |
|---|---|
| Molecular Formula | C2H3KO2 |
| Molecular Weight (g/mol) | 98.142 |
| MDL Number | MFCD00012458 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
| PubChem CID | 517044 |
| ChEBI | CHEBI:32029 |
| IUPAC Name | potassium;acetate |
| SMILES | CC(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC397121000
|
Thermo Scientific Chemicals
397121000 |
100 g | Glass bottle |
Each for $55.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for molecular biologyChemical Identifiers
| 127-08-2 | |
| 98.142 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
Specifications
| 292.0°C | |
| 99.0 | |
| White to Brown | |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.14 | |
| Molecular Biology | |
| >250°C | |
| Low Melting Solid |
| 127-08-2 | |
| 100.0 | |
| 99+% | |
| CH3CO2K | |
| 15, 7726 | |
| Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| ≥99% | |
| 100 g | |
| Glass bottle | |
| Acetic acid, potassium salt |
RUO – Research Use Only