missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific™

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446825000
View Documents
Thermo Scientific Chemicals
446825000
500g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC446820025
View Documents
Thermo Scientific Chemicals
446820025
2.5kg Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Chemical Identifiers

127-08-2
98.14
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]
C2H3KO2
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate

Specifications

127-08-2
99+%
C2H3KO2
500g
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14
Biochemistry
>250°C
Plastic bottle
CH3CO2K
15,7726
(5 M in water) Clear colorless solution
CC(=O)[O-].[K+]
98.14
CHEBI:32029
99+%
Acetic acid, potassium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-822-2

SDS
Documents

Documents

RUO – Research Use Only