missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for analysis

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

$118.04 - $118.04

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC217105000
SDS View Documents
Thermo Scientific Chemicals
217105000
500 g Plastic bottle
Each for $118.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

7.5 to 8.5 (5% soln. at 20°C)
127-08-2
100.0
99+%
CH3CO2K
02, 96
15, 7726
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
CC(=O)[O-].[K+]
98.142
CHEBI:32029
≥99%
500 g
Authentic
Granular Powder
292.0°C
99.0
White
C2H3KO2
MFCD00012458
01,906; 06,475
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14
Analytical
>250°C
Plastic bottle
Acetic acid, potassium salt, For analysis ACS
Videos
SDS
Documents

Documents

RUO – Research Use Only