missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, ACS reagent
CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
$150.19 - $150.19
Chemical Identifiers
| CAS | 127-08-2 |
|---|---|
| Molecular Formula | C2H3KO2 |
| Molecular Weight (g/mol) | 98.142 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
| PubChem CID | 517044 |
| ChEBI | CHEBI:32029 |
| IUPAC Name | potassium;acetate |
| SMILES | CC(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC418175000
|
Thermo Scientific Chemicals
418175000 |
500 g | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 127-08-2 | |
| 98.142 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
| C2H3KO2 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
Specifications
| 6.5 to 9.0 (5% soln. at 25°C) | |
| 127-08-2 | |
| 99+% | |
| CH3CO2K | |
| 01,906; 06,475 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.14 | |
| ACS Reagent | |
| >250°C | |
| Plastic bottle | |
| Acetic acid, potassium salt |
| 292.0°C | |
| White | |
| C2H3KO2 | |
| 02, 96 | |
| 15, 7726 | |
| Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| ≥99% | |
| 500 g | |
| Authentic | |
| Crystalline Powder |
RUO – Research Use Only