missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, ACS reagent

Catalog No. AC418175000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418175000 500 g Plastic bottle
AC418170025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418175000 Supplier Thermo Scientific Chemicals Supplier No. 418175000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

pH 6.5 to 9.0 (5% soln. at 25°C)
Melting Point 292.0°C
Color White
Quantity 500 g
Flash Point >250°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 99+%
Linear Formula CH3CO2K
Beilstein 02, 96
Fieser 01,906; 06,475
Merck Index 15, 7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less

RUO – Research Use Only