missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid HPLC, Lab Chem

Catalog No. LC100904
Change view
Click to view available options
Boiling Point:
118 °C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Boiling Point
LC100904 118 °C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC100904 Supplier LabChem Supplier No. LC100904

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Acetic Acid HPLC Acid Reagent, Safety: Corrosive, Acid

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.052
IUPAC Name Ethanolic Acid

Specifications

pH <1
Melting Point 16.7 °C
Boiling Point 118 °C
Color Colorless
Density 1.05 g/cm3
Quantity 4 L
Flash Point 39 °C
Ignition Point 427 °C
Odor Acrid
Packaging Amber Glass Bottle
Physical Form Liquid (solid below 17 °C)
Residue after Evaporation <5 ppm
Specific Gravity 1.0496
Linear Formula CH3COOH
Vapor Density 2.1
Vapor Pressure 11.4 mmHg
UN Number UN2789
Solubility Information Soluble
Formula Weight 60.052
Percent Purity >99.7%
Grade HPLC
Chemical Name or Material Acetic Acid Glacial
Show More Show Less
Hazard Category Corrosive
Hazard Statement Flammable
ShelfLife 5 years
DOTInformation UN2789
Recommended Storage Above 17 °C