missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (TraceMetal™ Grade), Fisher Chemical™

Catalog No. a507p212
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A507P212 2.5 L
A507P500 500 mL
A5074 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A507P212 Supplier Seastar Chemicals Inc Supplier No. S010601TFIQ09
Only null left
  • Certificate of Analysis included with each shipment.
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in nonbreakable polypropylene bottles to maintain product integrity and to
  • eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA. 45 mm bottle neck size

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Linear Formula CH3COOH
Formula Weight 60.05
Grade TraceMetal
Density 1.048g/cm³
Quantity 2.5 L
Packaging HDPE Bottle
Physical Form Liquid
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
DOTInformation DOT Class 8, : Corrosive