missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, TraceGrade, 99%, Spectrum™ Chemical

Catalog No. 18600233
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600233 500 mL Poly Bottle
18600235 2.5 L Poly Bottle
18600234 6 x 500 mL Poly Bottle
18600236 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600233 Supplier Spectrum Chemical Mfg Cor Supplier No. A1008500ML

A1008, 64-19-7, C2H4O2

Spectrum™ Chemical Acetic Acid, Glacial, TraceGrade also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak acid, when concentrated it is corrosive and can attack the skin. Spectrum’s TraceGrade products are suitable for use trace metal analysis and provides the lowest levels of metals, organics, and particulates available; giving you improved analytical performance and minimizing repeated runs.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 100%
UN Number UN2789
Percent Purity 99%
Grade High Purity
Quantity 500 mL
Packaging Poly Bottle