missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, Purified, Reagents

Catalog No. 01336858
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336858 2.5 L
01336857 20 L
01336856 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336858 Supplier Labchem Inc Supplier No. C51005002.5D1
Only null left

Acetic Acid, Glacial, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Boiling Point 118°C
Color Colorless
Grade Pure
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.0496
Chemical Name or Material Acetic Acid, Glacial, Purified