missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial (Optima™), Fisher Chemical™

Catalog No. p-20053
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A465250 250 mL FEP Bottle
A465500 500 mL FEP Bottle
A4651 1 L FEP Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A465250 Supplier Seastar Chemicals Inc Supplier No. S020601TFIF01
Only null left
  • Certificate of Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Linear Formula CH3COOH
UN Number 2789
Merck Index 15, 54
Formula Weight 60.05
Grade OPTIMA
Density 1.048 g/cm⊃3
Quantity 250 mL
Packaging FEP Bottle
Physical Form Liquid
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive