missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7156900
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600207 100 mL Amber Glass Bottle
18600208 500 mL Amber Glass Bottle
18600210 2.5 L Poly-Coated Glass Bottle
18600209 6 x 500 mL Amber Glass Bottle
18600211 4 L Poly-Coated Glass Bottle
18600212 4 x 2.5 L Poly-Coated Glass Bottle
18600213 4 x 4 L Poly-Coated Glass Bottle
18600214 20 L Poly Drum
18600215 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18600207 Supplier Spectrum Chemical Mfg Cor Supplier No. A1009100ML
Only null left

A1009, 64-19-7, C2H4O2

Spectrum™ Chemical Acetic Acid, Glacial, FCC is used in the pickling of vegetables and other foods. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 100%
UN Number UN2789
Percent Purity 99.5 to 100.5%
Grade FCC
Quantity 100 mL
Packaging Amber Glass Bottle