missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, FCC, 99.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Acetic Acid, Glacial, FCC is used in the pickling of vegetables and other foods. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| UN Number | UN2789 |
| Percent Purity | 99.5 to 100.5% |
| Grade | FCC |
| Quantity | 4 x 4 L |
| Packaging | Poly-Coated Glass Bottle |