missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, HPLC grade, MilliporeSigma™

Catalog No. MAX00746
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glas bottle (Pre packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX00746 500 mL Glas bottle (Pre packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX00746 Supplier Technidata Supplier No. AX00746

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 17°C
Boiling Point 116°C to 118°C (1013hPa)
Color Colorless
Quantity 500 mL
Flash Point 39°C
Odor Stinging
Packaging Glas bottle (Pre packed)
Physical Form Liquid
Residue after Evaporation 5ppm
Vapor Pressure 15.4 hPa (20°C)
Solubility Information 602.9 g/L
Formula Weight 60.05
Percent Purity ≤99.7%
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H226 Flammable liquid and vapor. H290 May be corrosive to meta
Hazard Statement P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
P233 Keep container tightly closed.
P234 Keep only in original container.
P240 Ground/bond container and receiving equipment.
P241 Use explosion-proof electrical/ ventilating/ lighting/ equipment.
P242 Use only non-sparking tools.
P243 Take precautionary measures against static discharge.
P264 Wash skin thoroughly after handling.
P280 Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331 IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P303 + P361 + P353 IF ON SKIN (or hair): Remove/ Take off immediately all contaminated clothing.
Rinse skin with water/ shower.
P304 + P340 IF INHALED: Remove victim to fresh air and keep atrest in a position comfortable forbreathing.
P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P310 Immediately call a POISON CENTER or doctor/ physician.
P321 Specific treatment (see supplemental first aid instructions on this label).
P363 Wash contaminated clothing before reuse.
P370 + P378 In case of fire: Use dry sand, dry chemical or alcohol-resistant foam for extinction.
P390 Absorb spillage to prevent material damage.
P403 + P235 Store in a well-ventilated place. Keep cool.
P405 Store locked up.
P406 Store in corrosive resistant stainless steel container with a resistant inner liner.
P501 Dispose of contents/ container to an approved waste disposal plant
EINECSNumber 200-580-7
RTECSNumber AF1225000