missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial (Certified ACS Plus), Fisher BioReagents
Description
Acetic Acid, Glacial can be used for several biochemical procedures. Some products are packed in Safe Cote™ which give additional safety for chemical and user both.
Specifications
Specifications
| pH | 2.5 |
| Melting Point | 16°C |
| Boiling Point | 117°C |
| Color | Colorless |
| Density | 1.048g/cm³ |
| Quantity | 500 mL |
| Packaging | Glass |
| Physical Form | Liquid |
| Residue after Evaporation | 8ppm max. |
| Linear Formula | CH3COOH |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |
| Recommended Storage | RT |