missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, ACS Reagent, Reagents
Acetic Acid, Glacial, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C5100600200E
Specifications
| Acetic Acid, Glacial, ACS Reagent | |
| 64-19-7 | |
| Colorless | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| ACS | |
| Poly Drum | |
| 1.0496 |
| <2 | |
| 118°C | |
| C2H4O2 | |
| Ethanoic Acid, Ethylic Acid | |
| CC(O)=O | |
| 60.05 | |
| 200 L | |
| Liquid |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| Ethanoic Acid, Ethylic Acid | |
| CC(O)=O |