missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, ACS Reagent, Reagents
Specifications
Specifications
| pH | <2 |
| Boiling Point | 118°C |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.0496 |
| Grade | ACS |
| Chemical Name or Material | Acetic Acid, Glacial, ACS Reagent |