missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, ACS Reagent Grade, Ricca Chemical

Catalog No. RABA0010500
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABA0010500 500 mL Glass Bottle
RABA00101C 1 L Amber Glass Bottle
RABA001025D 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RABA0010500 Supplier Ricca Chemical Company Supplier No. RABA0010500D1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 98.0
Color Colorless
CAS Max % 100.0
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
Percent Purity 99.7% (min.)
Grade ACS
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
ShelfLife 60 months