Learn More
Acetic Acid, Glacial, ACS Grade, ≥ 99.7%, LabChem™
Supplier: LabChem LC101003S
Specifications
| Acetic Acid | |
| 17°C | |
| 118°C | |
| Colorless | |
| CH3COOH | |
| UN2789 | |
| Soluble in water | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ≥99.7% | |
| 1049kg/m3 | |
| 40°C | |
| PVC Coated | |
| ≤0.0010% |
| Glacial | |
| 64-19-7 | |
| 100 | |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05 | |
| ACS | |
| 2.5 L | |
| Vinegar | |
| Liquid | |
| 1g/mL |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/spark/open flames/hot surfaces.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state, and federal regulations.
Danger
EINECSNumber : 607-002-00-6
Recommended Storage : Room Temperature