missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, ACS Grade, ≥ 99.7%, LabChem™

Supplier:  LabChem LC1010036

 View more versions of this product

Catalog No. LC1010036

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
17°C
118°C
Colorless
CH3COOH
UN2789
Soluble in water
CC(O)=O
60.05
CHEBI:15366
≥99.7%
1049kg/m3
40°C
HDPE Bottle
≤0.0010%
Glacial
64-19-7
100
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
ACS
6 x 2.5 L
Vinegar
Liquid
1g/mL

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/spark/open flames/hot surfaces.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state, and federal regulations.

Danger

EINECSNumber : 607-002-00-6

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions