missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, glacial, ≥99%, MP Biomedicals™
Specifications
Specifications
| Assay Percent Range | ≥99% |
| Formula Weight | 60.05 |
| Density | 1.049g/cm3 at 20°C |
| Quantity | 500 g |
| Odor | Very Pungent |
| Physical Form | Liquid |
| Chemical Name or Material | Acetic acid, glacial |