missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, 99.9985% (metals basis)

Catalog No. AA10994AP
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10994AP 500 mL
AA10994AC 25 mL
AA10994AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10994AP Supplier Thermo Scientific Chemicals Supplier No. 010994AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

Acetic acid is used as a solvent for purification and production of many chemical compounds eg., vinyl acetate monomer. It is a significant chemical reagent and is mainly used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as a descaling agent in households. As a food additive, it finds application as a condiment, in the pickling of vegetables and other foods and is also an acidity regulator. In analytical chemistry, for the estimation of weakly alkaline substances such as organic amides, glacial acetic acid is used. Acetic acid is used as antimicrobial preservative or acidulates in a variety of food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16.6°C
Boiling Point 118.1°C
Density 1.049 g/mL
Quantity 500 mL
Flash Point 40°C (104°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.3721
UN Number UN2789
Merck Index 14,55
Solubility Information Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide.
Formula Weight 60.05
Percent Purity 99.9985%
Assay (metals basis)
Chemical Name or Material Acetic acid, Glacial
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL
EINECSNumber 200-580-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only