Learn More
Acetic acid, glacial, 99.9985% (metals basis)
Description
Acetic acid is used as a solvent for purification and production of many chemical compounds eg., vinyl acetate monomer. It is a significant chemical reagent and is mainly used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as a descaling agent in households. As a food additive, it finds application as a condiment, in the pickling of vegetables and other foods and is also an acidity regulator. In analytical chemistry, for the estimation of weakly alkaline substances such as organic amides, glacial acetic acid is used. Acetic acid is used as antimicrobial preservative or acidulates in a variety of food products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| pH | 2.5 |
| Melting Point | 16.6°C |
| Boiling Point | 118.1°C |
| Density | 1.049 g/mL |
| Quantity | 25 mL |
| Flash Point | 40°C (104°F) |
| Odor | Pungent |
| Physical Form | Liquid |
| Refractive Index | 1.3721 |
| UN Number | UN2789 |
| Show More |
Safety and Handling
| Hazard Category | H226-H314 |
| Hazard Statement | GHS H Statement H314-H226 Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL |
| EINECSNumber | 200-580-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only