missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, 99.0%, MilliporeSigma™

Catalog No. 50947872
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947872 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947872 Supplier Technidata Supplier No. AX00771

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7
Molecular Weight (g/mol) 60.05

Specifications

pH 2.5 (50 g/L, H2O, 20°C)
Melting Point 17°C
Boiling Point 116 to 118°C (1013 hPa)
Quantity 500 mL
Flash Point 39°C
Packaging Glass Bottle, Pre-Packed
Assay Percent Range 99% min
Vapor Pressure 15.4 hPa (20°C)
Solubility Information 602.9 g/L
Recommended Storage Store at room temperature