missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid (glacial), 100%, EMPROVE™ ESSENTIAL Ph Eur, BP, JP, USP, E 260, MilliporeSigma™

Catalog No. M1000562500
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1000562500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1000562500 Supplier MilliporeSigma Supplier No. 1.00056.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
Synonym Ethanoic acid

Specifications

pH 2.5 (50 g/L, H2O, 20°C)
Melting Point 17°C
Boiling Point 116 to 118°C (1013 hPa)
Density 1.05 g/cm3 (at 20°C)
Quantity 2.5 L
Explosion Limit 4 to 19.9% (V)
Flash Point 39°C
Ignition Point 485°C
Packaging Glass Bottle
Residue after Evaporation ≤ 0.005%
Assay Percent Range Alkalimetric: 99.8 to 100.5%
Vapor Pressure 15.4 hPa (20°C)
Solubility Information 602.9 g/L Soluble
Show More Show Less
Recommended Storage Store at 15 to 25°C