missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid concentrate, For 1L standard solution, 1.0 M CH3COOH (1N), Honeywell Fluka™

Catalog No. 6000015
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000015 1 Pc.
60046682 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000015 Supplier Honeywell Chemicals Supplier No. 380511EA
Only null left

for 1L standard solution, 1.0 M CH3COOH (1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Quantity 1 Pc.
Flash Point 40°C (104°F)
Packaging Ampoule
Chemical Name or Material Acetic acid concentrate
Hazard Statement P280-P305 + P351 + P338-P310