Learn More
Acetic Acid, Certified, 30.0% (v/v) ±1.0%, LabChem™
Supplier: LabChem LC102604
Specifications
| Acetic Acid | |
| 69.69,30.31 | |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05 | |
| Certified | |
| 4 L | |
| Poly Bottle | |
| 1.04g/mL |
| 64-19-7,7732-18-5 | |
| Colorless | |
| CH3COOH | |
| UN2790 | |
| Soluble in water | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| 30.0% ±1.0% (v/v) | |
| 1.04g/mL | |
| Vinegar | |
| Liquid | |
| Traceable to NIST |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature