missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 30.0% (v/v) ±1.0%, LabChem™

Supplier:  LabChem LC102602

 View more versions of this product

Catalog No. LC102602


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
69.69,30.31
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Certified
1 L
Poly Bottle
1.04g/mL
64-19-7,7732-18-5
Colorless
CH3COOH
UN2790
Soluble in water
CC(O)=O
60.05
CHEBI:15366
30.0% ±1.0% (v/v)
1.04g/mL
Vinegar
Liquid
Traceable to NIST

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions