missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 1.000N ±0.005N (1.0M), LabChem™

$43.80 - $383.97

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC103801
View Documents
LabChem
LC103801
500 mL
Each for $43.80
Only null left
 
LC103802
View Documents
LabChem
LC103802
1 L
Each for $66.00
Only null left
 
LC103804
View Documents
LabChem
LC103804
4 L
Each for $130.40
Only null left
 
LC103805
View Documents
LabChem
LC103805
20 L
Each for $383.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

64-19-7,7732-18-5
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
60.05
CHEBI:15366
Certified
500 mL
Poly Bottle
1.01g/mL
Acetic Acid
94.04,5.96
C2H4O2
MFCD00036152
Soluble in water
CC(O)=O
1.000N ±0.005N (1.0M)
176
60.05
1.01g/mL
Vinegar
Liquid
Traceable to NIST
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin iritation and serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'