missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid-Ammonium Acetate Buffer TS, Ricca Chemical
$186.10 - $352.23
Specifications
| Chemical Name or Material | Acetic Acid-Ammonium Acetate Buffer |
|---|---|
| Name Note | Test Solution |
| CAS | 7732-18-5,631-61-8,64-19-7 |
| CAS Min % | 86.0,6.65,5.16 |
| CAS Max % | 90.0,6.92,5.37 |
Specifications
| Acetic Acid-Ammonium Acetate Buffer | |
| 7732-18-5,631-61-8,64-19-7 | |
| 90.0,6.92,5.37 | |
| 1X | |
| 4.7 | |
| Buffer | |
| Laboratory |
| Test Solution | |
| 86.0,6.65,5.16 | |
| Colorless | |
| Product Monographs | |
| Liquid | |
| Natural Poly Bottle |