missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥99.7%

$506.28 - $506.28

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000005
View Documents
Honeywell-Fluka
695092500ML
500 mL
Each for $506.28
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

64-19-7
CH3CO2H
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
≥99.7%
1.049g/mL at 25°C
16% at 92°F, 4% at 59°F
800°F
11.4 mmHg (20°C)
C2H4O2
MFCD00036152
506007
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05g/mol
ACS Reagent
500 mL
40°C (104°F)
2.07 (vs air)
Acetic acid
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378

RTECSNumber : AF1225000

SDS
missing translation for 'documents'

missing translation for 'documents'