missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, 99+%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 16.0°C to 16.5°C |
| Boiling Point | 117.0°C to 118.0°C |
| Assay Percent Range | 99+% |
| Linear Formula | CH3COOH |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide |
| Formula Weight | 60.05 |
| Percent Purity | 99+% |
| Grade | Extra Pure |
| Density | 1.0480g/mL |
| Quantity | 5 L |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | Danger |
| missing translation for 'healthHazard2' | GHS P Statement Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only