missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, ≥99.99% (trace metals basis), Honeywell
≥99.99% trace metals basis
$108.00 - $933.13
Chemical Identifiers
| CAS | 64-19-7 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | 60.05 |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6000024
|
Honeywell Chemicals
33882625ML |
25 mL |
Each for $108.00
|
|
|||||
|
6000023
|
Honeywell Chemicals
338826100ML |
100 mL |
Each for $293.53
|
|
|||||
|
6000025
|
Honeywell Chemicals
3388264X100ML |
4 x 100 mL |
Case of 4 Each for $933.13
|
|
|||||
|
6000026
|
Honeywell Chemicals
338826500ML |
500 mL |
Each for $899.53
|
|
|||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| 64-19-7 | |
| CH3CO2H | |
| UN2789 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ≥99.99% (trace metals basis) | |
| 25 mL | |
| 40°C (104°F) | |
| 2.07 (vs air) |
| C2H4O2 | |
| MFCD00036152 | |
| 506007 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05g/mol | |
| 1.049g/mL at 25°C | |
| 4% at 59°F, 16% at 92°F | |
| 800°F | |
| Acetic acid |
Safety and Handling
P280-P305 + P351 + P338-P310
RTECSNumber : AF1225000