Learn More
Acetic Acid, 99.8%, for analysis
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Supplier: Thermo Scientific Chemicals 222140200
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
| Acetic acid | |
| 16.0°C to 16.5°C | |
| 117.0°C to 118.0°C | |
| 100.0 | |
| C2H4O2 | |
| MFCD00036152 | |
| 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05 | |
| Analytical | |
| 20 L | |
| Plastic Can | |
| 1.3700 to 1.3730 | |
| 1.048 |
| For Analysis | |
| 64-19-7 | |
| 99.75 | |
| 99.8% | |
| CH3COOH | |
| 02, 96 | |
| 15, 54 | |
| Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| 99.8% | |
| 1.0480g/mL | |
| 40°C | |
| Liquid | |
| 0.001% max. |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
RUO – Research Use Only