missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 25%, solution in water

Catalog No. AC456850250
Change view
Click to view available options
Quantity:
5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456850250 25 L
AC456850050 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456850250 Supplier Thermo Scientific Chemicals Supplier No. 456850250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-19-7 | C2H4O2 | 60.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point -17.0°C
Boiling Point 103.0°C
Density 1.0400g/mL
Quantity 25 L
Flash Point >100°C
Specific Gravity 1.04
Linear Formula CH3CO2H
Solubility Information Solubility in water: completely soluble.
Formula Weight 60.05
Percent Purity 25%
Chemical Name or Material Acetic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

RUO – Research Use Only