missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 2.00 N, Ricca Chemical
$79.09 - $136.16
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| 7732-18-5,64-19-7 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| acetic acid | |
| 176 | |
| Laboratory | |
| Natural Poly Bottle | |
| Acetic Acid |
| 87.0,10.51 | |
| 91.0,10.94 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 2.00 Normal | |
| Mixture | |
| CHEBI:15366 | |
| 500 mL | |
| Liquid |