missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid (1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenylpropyl Ester 98.0+%, TCI America™

Catalog No. A15341G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15341G Supplier TCI America Supplier No. A15341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Reagent for double aldol reaction]

Chemical Identifiers

CAS 240423-74-9
Molecular Formula C27H31NO4S
Molecular Weight (g/mol) 465.61
MDL Number MFCD02093424
InChI Key MCELVTKIIIBSDU-UHFFFAOYNA-N
Synonym (1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)-O-acetylnorephedrine, (1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenylpropyl Acetate
PubChem CID 44630064
IUPAC Name 2-(N-benzyl2,4,6-trimethylbenzenesulfonamido)-1-phenylpropyl acetate
SMILES CC(C(OC(C)=O)C1=CC=CC=C1)N(CC1=CC=CC=C1)S(=O)(=O)C1=C(C)C=C(C)C=C1C

Specifications

Melting Point 121°C
Color White
Quantity 1 g
Formula Weight 465.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acetic Acid (1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenylpropyl Ester [Reagent for doubl