missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 1.00 N, Ricca Chemical
Shelf Life: 18 months
$85.28 - $584.25
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
15016
|
Ricca Chemical Company
15016 |
500 mL | Natural Poly Bottle |
Each for $85.28
|
|
||||
|
150-32
|
Ricca Chemical Company
RI15032 |
1 L | Natural Poly Bottle |
Each for $102.53
|
|
||||
|
150-1
|
Ricca Chemical Company
1501 |
4 L | Natural Poly Bottle |
Each for $238.91
|
|
||||
|
150-5
|
Ricca Chemical Company
RI1505 |
20 L | Cubitainer |
Each for $584.25
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| 7732-18-5,64-19-7 | |
| 96.0,5.79 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| Laboratory | |
| Natural Poly Bottle |
| 92.0,5.56 | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| Mixture | |
| CHEBI:15366 | |
| 500 mL | |
| Acetic Acid 1N |