missing translation for 'onlineSavingsMsg'
Learn More

Acéthydrazide, 95%

Numéro de catalogue. AC102350250
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Bouteille en verre
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC102350250 25 g Bouteille en verre
AC102351000 100 g Bouteille en verre
AC102355000 500 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC102350250 Fournisseur Thermo Scientific Chemicals Code fournisseur 102350250
Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

CAS : 1068-57-1 | C2H 6N 2O | 74,08 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1068-57-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00007610
InChI Key OFLXLNCGODUUOT-UHFFFAOYSA-N
Synonym acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
PubChem CID 14039
ChEBI CHEBI:48978
IUPAC Name Acétohydrazide
SMILES CC(=O)NN

Spécifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color Blanc
Boiling Point 129.0°C (18.0 mmHg)
Flash Point >110°C
Loss on Drying Balance (50°C, 3 Hrs)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance)
Packaging Bouteille en verre
Linear Formula CH3CONHNH2
Quantity 25 g
Beilstein 02,191
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,chloroform
Formula Weight 74.08
Percent Purity 95%
Physical Form Cristaux humides et morceaux
Chemical Name or Material Acethydrazide, 95%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Flammable solid.
Risk of explosion if heated under confinement.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-948-7
RTECSNumber AI1225000
TSCA TSCA

RUO – Usage réservé à la recherche