missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012219
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012219 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012219 Supplier Merck Emd Millipore Supplier No. PHR19085G
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 59-66-5
Linear Formula C4H6N4O3S2
MDL Number MFCD00003105
Synonym Acetazolamide; 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide; N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide; N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide
Type Certified Reference Material
Quantity 5 g
Formula Weight 222.25
Grade Certified Reference Material