missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS | 127-09-3 |
| Quantity | 1 L |
| Molecular Formula | C2H3NaO2 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| SMILES | [Na+].CC([O-])=O |
| IUPAC Name | sodium acetate |
| Molecular Weight (g/mol) | 82.03 |