missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer Solution, pH 3.5, for USP Heavy Metals Testing, Ricca Chemical
$192.79 - $859.70
Specifications
| Chemical Name or Material | Acetate Buffer Solution |
|---|---|
| pH | 3.5 |
| Physical Form | Liquid |
| Solution Type | Buffer |
| Packaging | Natural Poly Bottle |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0060100120
|
Ricca Chemical Company
R0060100120A |
120 mL |
Each for $192.79
|
|
|||||
|
60132
|
Ricca Chemical Company
60132 |
1 L |
Each for $859.70
|
|
|||||
Specifications
| Acetate Buffer Solution | |
| Liquid | |
| Natural Poly Bottle |
| 3.5 | |
| Buffer | |
| Laboratory |