missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer pH 6.0, For Aluminum, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Acetate Buffer pH 6.0 |
| Name Note | For Aluminum |
| CAS | 7732-18-5, 6131-90-4, 64-19-7 |
| Color | Colorless |
| CAS Max % | 86.17, 13.6, 0.23 |
| Decomposition Information | Carbon monoxide, Carbon dioxide |
| For Use With (Application) | Aluminum Analysis |
| pH | 6.00 ±0.05 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Show More |