missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 6.0 EP Buffer Solution 4002200, Reagents

Catalog No. 01336018
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336018 1 L
01336017 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336018 Supplier Labchem Inc Supplier No. CA0155001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetate Buffer, pH 6.0 EP Buffer Solution 4002200, Reagent, use for European pharmacopeia. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetate Buffer, pH 6.0 EP Buffer Solution 4002200
CAS 7732-18-5, 631-61-8, 631-61-8
Boiling Point 100°C
Color Colorless
pH 6
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.1
Sterility Non-sterile
Grade Reagent
Show More Show Less