missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 5.5, Ricca Chemical

Catalog No. R00650001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00650001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00650001A Supplier Ricca Chemical Company Supplier No. R00650001A
Only null left

Specifications

Chemical Name or Material Acetate Buffer
CAS 64-19-7, 7732-18-5, 1336-21-6
CAS Max % 48.15, 43.76, 8.09
Color Colorless Liquid
Concentration 1X
For Use With (Application) Water Analysis
pH 5.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Laboratory
Show More Show Less