missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 4, Reagents

Catalog No. 01336151
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336151 10 L
01336150 1 L
01336149 20 L
01336148 4 L
01336147 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336151 Supplier Labchem Inc Supplier No. CA00290010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetate Buffer, pH 4, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetate Buffer, pH 4
CAS 7732-18-5, 64-19-7, 64-19-7
Boiling Point 100°C
Color Colorless
pH 4
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Packaging Cubitainer
Specific Gravity 1.0627
Sterility Non-sterile
Grade Reagent
Show More Show Less