missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 4.6, for Bromide Analysis, Ricca Chemical

Catalog No. 5416
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5416 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5416 Supplier Ricca Chemical Company Supplier No. 5416

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Analysis of Bromide

Specifications

Chemical Name or Material Acetate Buffer
Name Note pH 4.6
CAS 7732-18-5, 7647-14-5, 6131-90-4, 64-19-7
CAS Min % 82.0, 7.42, 5.61, 2.59
Color Colorless
CAS Max % 86.0, 7.72, 5.83, 2.7
Concentration 1X
For Use With (Application) Water Analysis
pH 4.6
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less